Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Btk
Entrez Gene ID
12229
Gene Name
Bruton agammaglobulinemia tyrosine kinase
Gene Aliases
AI528679, xid
Chromosome Location
Chr.X: 134542334 - 134583623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013482.2 NP_038510.2 6 - 7 680 154
GenBank mRNA BC053392.1 - 6 - 7 616 154
DT921900.1 - 2 - 3 155
L08967.1 - 6 - 7 656 154
L10627.1 - 6 - 7 685 160
L29788.1 - 6 - 7 652 154

View Full Product Details