Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd3d
Entrez Gene ID
12500
Gene Name
CD3 antigen, delta polypeptide
Gene Aliases
T3d
Chromosome Location
Chr.9: 44981769 - 44987052 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013487.3 NP_038515.3 1 - 2 160 100
GenBank mRNA AK042172.1 - 1 - 2 162 100
AK050629.1 - 1 - 2 149 100
AK153843.1 - 1 - 2 125 100
AK154064.1 - 1 - 2 122 100
BC062807.1 - 1 - 2 100
EF382667.1 - 1 - 2 60 100
HM536173.1 - 1 - 2 60 100

View Full Product Details