Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Myh11
Entrez Gene ID
17880
Gene Name
myosin, heavy polypeptide 11, smooth muscle
Gene Aliases
AV071570, SM1, SM2, smMHC
Chromosome Location
Chr.16: 14194527 - 14291408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161775.1 NP_001155247.1 7 - 8 943 91
NM_013607.2 NP_038635.2 7 - 8 943 91
GenBank mRNA AK137226.1 - 8 - 9 921 91
AK137716.1 - 7 - 8 899 91
AK173045.1 - 7 - 8 907 91
BC026142.1 - 7 - 8 928 91
D85923.1 - 7 - 8 898 91
D85924.1 - 7 - 8 898 91
L25860.1 - 7 - 8 887 91

View Full Product Details