Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Myh11
Entrez Gene ID
17880
Gene Name
myosin, heavy polypeptide 11, smooth muscle
Gene Aliases
AV071570, SM1, SM2, smMHC
Chromosome Location
Chr.16: 14194527 - 14291408 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161775.1 NP_001155247.1 36 - 37 5323 60
NM_013607.2 NP_038635.2 36 - 37 5323 60
GenBank mRNA AK173045.1 - 36 - 37 5287 60
BC026142.1 - 36 - 37 5308 60
D85923.1 - 36 - 37 5278 60
D85924.1 - 36 - 37 5278 60

View Full Product Details