The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013665.1 | NP_038693.1 | 1 - 2 | 560 | 97 |
GenBank mRNA | AK032007.1 | - | 1 - 2 | 107 | 97 |
U66918.1 | - | 1 - 2 | 560 | 97 |