Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tek
Entrez Gene ID
21687
Gene Name
endothelial-specific receptor tyrosine kinase
Gene Aliases
AA517024, Cd202b, Hyk, STK1, Tie-2, Tie2
Chromosome Location
Chr.4: 94739289 - 94874976 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290549.1 NP_001277478.1 18 - 19 3338 75
NM_001290551.1 NP_001277480.1 17 - 18 3188 75
NM_013690.3 NP_038718.2 18 - 19 3341 75
GenBank mRNA BC050824.1 - 17 - 18 3182 75
D13738.1 - 18 - 19 3346 75
S67051.1 - 19 - 20 3001 75
X67553.1 - 18 - 19 3109 75
X71426.1 - 19 - 20 3072 75

View Full Product Details