The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013865.2 | NP_038893.1 | 5 - 6 | 477 | 74 |
NM_180956.1 | NP_851287.1 | 5 - 6 | 477 | 74 | |
GenBank mRNA | AB033922.1 | - | 5 - 6 | 391 | 74 |
AK002272.1 | - | 4 - 5 | 391 | 74 | |
AK004689.1 | - | 5 - 6 | 423 | 74 | |
AK036277.1 | - | 5 - 6 | 402 | 74 | |
AK037487.1 | - | 5 - 6 | 403 | 74 | |
AK154927.1 | - | 5 - 6 | 444 | 74 | |
AK209466.1 | - | 2 - 3 | 191 | 74 | |
AK214669.1 | - | 1 - 2 | 47 | 74 | |
BC018504.1 | - | 5 - 6 | 411 | 74 |