Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp11a
Entrez Gene ID
50770
Gene Name
ATPase, class VI, type 11A
Gene Aliases
4930558F19Rik, AU040868, Atpc1h, Ih
Chromosome Location
Chr.8: 12757016 - 12868728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293667.1 NP_001280596.1 18 - 19 2299 68
NM_001293668.1 NP_001280597.1 18 - 19 2299 68
NM_015804.3 NP_056619.1 18 - 19 2299 68
NR_121603.1 18 - 19 2299 68
GenBank mRNA AF156551.1 - 18 - 19 2091 68
AK147497.1 - 18 - 19 2302 68
AK173088.1 - 18 - 19 2452 68
BC036322.1 - 3 - 4 360 68
BC138715.1 - 18 - 19 2018 68
BC145399.1 - 18 - 19 2019 68
BC145400.1 - 18 - 19 2019 68

View Full Product Details