The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122982.2 | NP_001116454.1 | 9 - 10 | 784 | 79 |
NM_001290420.1 | NP_001277349.1 | 9 - 10 | 784 | 79 | |
NM_001290422.1 | NP_001277351.1 | 9 - 10 | 784 | 79 | |
NM_016746.4 | NP_058026.2 | 9 - 10 | 784 | 79 | |
GenBank mRNA | AK009615.1 | - | 9 - 10 | 730 | 79 |
AK014079.1 | - | 8 - 9 | 633 | 79 | |
AK037957.1 | - | 9 - 10 | 785 | 79 | |
AK135472.1 | - | 9 - 10 | 780 | 79 | |
AK153035.1 | - | 9 - 10 | 717 | 79 | |
BC003344.1 | - | 9 - 10 | 717 | 79 | |
BC029211.1 | - | 9 - 10 | 726 | 79 | |
BC037689.1 | - | 9 - 10 | 712 | 79 | |
BC062376.1 | - | 9 - 10 | 699 | 79 | |
BC087544.1 | - | 9 - 10 | 716 | 79 | |
BC100396.1 | - | 9 - 10 | 758 | 79 | |
BC120649.1 | - | 9 - 10 | 711 | 79 | |
BC120677.1 | - | 9 - 10 | 711 | 79 | |
BQ175960.1 | - | 3 - 4 | 79 | ||
U62638.1 | - | 9 - 10 | 759 | 79 |