Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Corin
Entrez Gene ID
53419
Gene Name
corin
Gene Aliases
AV273130, Lrp4
Chromosome Location
Chr.5: 72299948 - 72504540 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122756.1 NP_001116228.1 20 - 21 2923 102
NM_016869.3 NP_058565.2 20 - 21 3080 102
GenBank mRNA AB013874.1 - 20 - 21 3080 102
BC093485.1 - 20 - 21 2856 102
BC138339.1 - 20 - 21 3063 102
BC138340.1 - 20 - 21 3063 102
BC145243.1 - 17 - 18 2724 102

View Full Product Details