The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016905.2 | NP_058601.2 | 5 - 6 | 834 | 58 |
GenBank mRNA | AB027012.1 | - | 5 - 6 | 790 | 58 |
AK010340.1 | - | 5 - 6 | 831 | 58 | |
AK011103.1 | - | 5 - 6 | 835 | 58 | |
AK045694.1 | - | 5 - 6 | 808 | 58 | |
AK088151.1 | - | 5 - 6 | 835 | 58 | |
AK168716.1 | - | 5 - 6 | 794 | 58 | |
BC016602.1 | - | 5 - 6 | 834 | 58 | |
BC050151.1 | - | 5 - 6 | 805 | 58 |