The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016923.2 | NP_058619.1 | 2 - 3 | 253 | 100 |
GenBank mRNA | AB018550.1 | - | 2 - 3 | 253 | 100 |
AF168120.1 | - | 2 - 3 | 223 | 100 | |
AK019283.1 | - | 2 - 3 | 277 | 100 | |
BC116785.1 | - | 2 - 3 | 249 | 100 |