Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgef7
Entrez Gene ID
54126
Gene Name
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location
Chr.8: 11728105 - 11835219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113517.1 NP_001106989.1 11 - 12 1415 107
NM_001113518.1 NP_001106990.1 10 - 11 1204 107
NM_017402.4 NP_059098.2 10 - 11 1204 107
GenBank mRNA AF247654.1 - 10 - 11 1100 107
AF247655.1 - 8 - 9 821 107
AF343877.1 - 10 - 11 1084 107
AK052545.1 - 10 - 11 1175 107
AK129064.1 - 11 - 12 1351 107
AK139156.1 - 11 - 12 1344 107
AY220301.1 - 10 - 11 1169 107
BC044838.1 - 10 - 11 1189 107
U96634.1 - 8 - 9 894 107

View Full Product Details