The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019454.3 | NP_062327.2 | 8 - 9 | 1600 | 71 |
GenBank mRNA | AB043893.1 | - | 8 - 9 | 1243 | 71 |
AF253469.1 | - | 8 - 9 | 1555 | 71 | |
AF273454.1 | - | 8 - 9 | 1621 | 71 | |
AK004739.1 | - | 8 - 9 | 1568 | 71 | |
AK031699.1 | - | 8 - 9 | 1570 | 71 | |
AK052322.1 | - | 8 - 9 | 1601 | 71 | |
AK165517.1 | - | 8 - 9 | 1568 | 71 | |
AK171717.1 | - | 8 - 9 | 1567 | 71 | |
BC042497.1 | - | 8 - 9 | 1555 | 71 | |
BC049130.1 | - | 8 - 9 | 1560 | 71 |