Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uso1
Entrez Gene ID
56041
Gene Name
USO1 vesicle docking factor
Gene Aliases
115kDa, TAP, Vdp
Chromosome Location
Chr.5: 92137909 - 92202800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019490.1 NP_062363.1 15 - 16 1836 105
GenBank mRNA AF096868.1 - 14 - 15 1624 105
AK156070.1 - 15 - 16 1829 105
AK168433.1 - 3 - 4 367 105
BC016069.1 - 15 - 16 1836 105

View Full Product Details