Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Git2
Entrez Gene ID
26431
Gene Name
G protein-coupled receptor kinase-interactor 2
Gene Aliases
1500036H07Rik, 5830420E16Rik, 6430510B20Rik, 9630056M03Rik, B230104M05Rik, CAT2, mKIAA0148
Chromosome Location
Chr.5: 114727408 - 114773520 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077359.1 NP_001070827.1 2 - 3 310 85
NM_001077360.1 NP_001070828.1 2 - 3 309 85
NM_019834.3 NP_062808.3 2 - 3 309 85
GenBank mRNA AF148693.1 - 2 - 3 183 85
AK017943.1 - 2 - 3 286 85
AK220323.1 - 2 - 3 310 85
BC043062.1 - 2 - 3 309 85
BC056993.1 - 2 - 3 309 85

View Full Product Details