Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pik3cg
Entrez Gene ID
30955
Gene Name
phosphoinositide-3-kinase, catalytic, gamma polypeptide
Gene Aliases
5830428L06Rik, PI3Kgamma, p110gamma
Chromosome Location
Chr.12: 32173129 - 32208677 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146200.1 NP_001139672.1 9 - 10 3011 73
NM_001146201.1 NP_001139673.1 9 - 10 3067 73
NM_020272.2 NP_064668.2 9 - 10 3188 73
GenBank mRNA AF208345.1 - 8 - 9 2867 73
AJ249280.1 - 9 - 10 3188 73
AK040543.1 - 9 - 10 2973 73
AK077772.1 - 3 - 4 343 73
AK132553.1 - 9 - 10 3013 73
AK137470.1 - 9 - 10 2920 73
AK137620.1 - 9 - 10 3069 73
AK143238.1 - 6 - 7 692 73
BC051246.1 - 9 - 10 2928 73

View Full Product Details