The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290821.1 | NP_001277750.1 | 6 - 7 | 1166 | 72 |
NM_020492.4 | NP_065238.2 | 6 - 7 | 1166 | 72 | |
GenBank mRNA | AK078258.1 | - | 6 - 7 | 1164 | 72 |
AY129229.1 | - | 6 - 7 | 713 | 72 | |
BC131949.1 | - | 6 - 7 | 831 | 72 | |
BC137725.1 | - | 6 - 7 | 831 | 72 | |
S73717.1 | - | 6 - 7 | 758 | 72 | |
S73718.1 | - | 6 - 7 | 758 | 72 |