Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kcne3
Entrez Gene ID
57442
Gene Name
potassium voltage-gated channel, Isk-related subfamily, gene 3
Gene Aliases
2210017H05Rik, MiRP2
Chromosome Location
Chr.7: 100176507 - 100184869 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190869.1 NP_001177798.1 4 - 5 602 97
NM_001190871.2 NP_001177800.1 3 - 4 367 97
NM_020574.5 NP_065599.1 3 - 4 301 97
GenBank mRNA AF076532.1 - 2 - 3 202 97
AK008744.1 - 3 - 4 290 97
AK132773.1 - 4 - 5 594 97
BC004629.1 - 3 - 4 333 97
DV056972.1 - 3 - 4 347 97
EH094997.1 - 1 - 2 97

View Full Product Details