The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021297.2 | NP_067272.1 | 2 - 3 | 276 | 117 |
GenBank mRNA | AF095353.1 | - | 2 - 3 | 462 | 117 |
AF110133.1 | - | 2 - 3 | 276 | 117 | |
AF185285.1 | - | 2 - 3 | 255 | 117 | |
AK030348.1 | - | 2 - 3 | 476 | 117 | |
AK154297.1 | - | 2 - 3 | 369 | 117 | |
AK170566.1 | - | 2 - 3 | 327 | 117 | |
BC029856.1 | - | 2 - 3 | 488 | 117 | |
JX878358.1 | - | 2 - 3 | 270 | 117 | |
JX878359.1 | - | 2 - 3 | 270 | 117 |