The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083904.1 | NP_001077373.1 | 6 - 7 | 849 | 83 |
NM_001083905.1 | NP_001077374.1 | 7 - 8 | 1019 | 83 | |
NM_021564.2 | NP_067539.1 | 7 - 8 | 1084 | 83 | |
GenBank mRNA | AJ242927.1 | - | 7 - 8 | 1005 | 83 |
AK009310.1 | - | 7 - 8 | 1074 | 83 | |
AK009554.1 | - | 7 - 8 | 1018 | 83 | |
AK037043.1 | - | 7 - 8 | 1020 | 83 | |
AY138459.1 | - | 7 - 8 | 1069 | 83 | |
AY138460.1 | - | 6 - 7 | 619 | 83 | |
BC018341.1 | - | 7 - 8 | 949 | 83 |