The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023126.2 | NP_075615.2 | 4 - 5 | 488 | 108 |
GenBank mRNA | AB232600.1 | - | 4 - 5 | 329 | 108 |
AK009259.1 | - | 4 - 5 | 339 | 108 | |
AK010862.1 | - | 4 - 5 | 339 | 108 | |
AK076048.1 | - | 4 - 5 | 367 | 108 | |
AK079306.1 | - | - | 355 | 108 | |
AK080740.1 | - | 4 - 5 | 340 | 108 | |
AK147178.1 | - | 4 - 5 | 356 | 108 | |
AK165690.1 | - | 4 - 5 | 351 | 108 | |
AY974570.1 | - | 4 - 5 | 307 | 108 | |
BC019990.1 | - | 4 - 5 | 329 | 108 | |
S53270.1 | - | 4 - 5 | 346 | 108 |