Product Details

Assay Reference Genome
Location

Chr.16:14448446 on build GRCm38
Cytoband
16A1
Species
Mus musculus
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
17250
Gene Symbol
Abcc1
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 1
Gene Aliases
Abcc1a, Abcc1b, MRP, Mdrap, Mrp1
Location
Chr.16:14361521-14474879 on build GRCm38
Assay Gene Location
Within Exon 19
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
Abcc1 NM_008576.3 19 2545 NP_032602.1
AF022908.1 19 2507 AAB80938.1
AK029876.1 19 2546 BAC26654.1
AK049174.1 3 312 BAC33586.1
BC090617.1 19 2516

View Full Product Details