The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190490.1 | NP_001177419.1 | 7 - 8 | 1029 | 96 |
NM_001190491.1 | NP_001177420.1 | 7 - 8 | 1029 | 96 | |
NM_032418.2 | NP_115794.1 | 7 - 8 | 1029 | 96 | |
GenBank mRNA | BC024150.1 | - | 7 - 8 | 1016 | 96 |
BC056615.1 | - | 7 - 8 | 1008 | 96 | |
BC075715.1 | - | 7 - 8 | 937 | 96 | |
S60313.1 | - | 4 - 5 | 536 | 96 | |
S60314.1 | - | 4 - 5 | 536 | 96 | |
S60315.1 | - | 4 - 5 | 536 | 96 | |
S60316.1 | - | 4 - 5 | 536 | 96 |