The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_007503.3 | NP_031529.2 | 3 - 4 | 206 | 96 |
NM_052823.2 | NP_439888.1 | 3 - 4 | 279 | 96 | |
NR_028499.1 | 3 - 4 | 411 | 96 | ||
GenBank mRNA | AA396010.1 | - | 3 - 4 | 227 | 96 |
AI316264.1 | - | 3 - 4 | 190 | 96 | |
AK002215.1 | - | 3 - 4 | 194 | 96 | |
AK002228.1 | - | 3 - 4 | 207 | 96 | |
AK007105.1 | - | 1 - 2 | 81 | 96 | |
AY626243.1 | - | 3 - 4 | 328 | 96 | |
AY626244.1 | - | 3 - 4 | 279 | 96 | |
AY626245.1 | - | 3 - 4 | 364 | 96 | |
BC024614.1 | - | 3 - 4 | 195 | 96 | |
BU756853.1 | - | 3 - 4 | 96 | ||
CB600439.1 | - | 3 - 4 | 239 | 96 | |
CB600958.1 | - | 3 - 4 | 414 | 96 | |
X70060.1 | - | 2 - 3 | 117 | 96 |