The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178589.3 | NP_848704.1 | 1 - 2 | 537 | 59 |
GenBank mRNA | AF322069.1 | - | 1 - 2 | 95 | 59 |
AK033529.1 | - | 1 - 2 | 514 | 59 | |
AK043823.1 | - | 1 - 2 | 539 | 59 | |
AK134704.1 | - | 1 - 2 | 508 | 59 | |
AK143175.1 | - | 1 - 2 | 524 | 59 | |
AY043489.1 | - | 1 - 2 | 95 | 59 | |
BC016420.1 | - | 1 - 2 | 486 | 59 |