The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053195.2 | NP_444425.1 | 16 - 17 | 1694 | 74 |
GenBank mRNA | AF314821.1 | - | 16 - 17 | 1734 | 74 |
BC005742.1 | - | 6 - 7 | 824 | 74 | |
BC017615.1 | - | 16 - 17 | 1694 | 74 |