The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_152804.2 | NP_690017.2 | 5 - 6 | 835 | 96 |
GenBank mRNA | AK143452.1 | - | 4 - 5 | 1410 | 96 |
AK164365.1 | - | 5 - 6 | 834 | 96 | |
BC034513.1 | - | 5 - 6 | 817 | 96 | |
BC035451.1 | - | 5 - 6 | 817 | 96 | |
M96163.1 | - | 5 - 6 | 801 | 96 |