Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptgis
Entrez Gene ID
19223
Gene Name
prostaglandin I2 (prostacyclin) synthase
Gene Aliases
Cyp8, Cyp8a1, Pgis
Chromosome Location
Chr.2: 167203194 - 167252127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008968.3 NP_032994.1 8 - 9 1326 71
GenBank mRNA AB001607.1 - 8 - 9 1244 71
AK048059.1 - - 1379 71
AK076585.1 - 8 - 9 1327 71
BC062151.1 - 8 - 9 1333 71

View Full Product Details