The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278605.1 | NP_001265534.1 | 2 - 3 | 228 | 93 |
NM_009470.5 | NP_033496.1 | 2 - 3 | 284 | 93 | |
GenBank mRNA | AK085460.1 | - | 2 - 3 | 278 | 93 |
AK144065.1 | - | 2 - 3 | 200 | 93 | |
AK164688.1 | - | 2 - 3 | 200 | 93 | |
AK165507.1 | - | 2 - 3 | 144 | 93 | |
BB843740.1 | - | 2 - 3 | 229 | 93 | |
BC012973.1 | - | 1 - 2 | 122 | 93 |