Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Umod
Entrez Gene ID
22242
Gene Name
uromodulin
Gene Aliases
THP, Urehd1, urehr4
Chromosome Location
Chr.7: 119462708 - 119479262 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278605.1 NP_001265534.1 8 - 9 1889 129
NM_009470.5 NP_033496.1 8 - 9 1945 129
GenBank mRNA AK085460.1 - 8 - 9 1939 129
AK144065.1 - 8 - 9 1861 129
AK164688.1 - 8 - 9 1861 129
AK165507.1 - 8 - 9 1805 129
BC012973.1 - 7 - 8 1783 129
L33406.1 - 8 - 9 1875 129

View Full Product Details