The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159401.1 | NP_001152873.1 | 8 - 9 | 1051 | 96 |
NM_001159402.1 | NP_001152874.1 | 8 - 9 | 1050 | 96 | |
NM_009477.3 | NP_033503.2 | 8 - 9 | 1078 | 96 | |
GenBank mRNA | AK136474.1 | - | 8 - 9 | 931 | 96 |
BC036559.1 | - | 8 - 9 | 952 | 96 | |
BX527866.1 | - | 2 - 3 | 295 | 96 | |
D44464.1 | - | 8 - 9 | 1079 | 96 |