Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mog
Entrez Gene ID
17441
Gene Name
myelin oligodendrocyte glycoprotein
Gene Aliases
B230317G11Rik
Chromosome Location
Chr.17: 37010740 - 37023398 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010814.2 NP_034944.2 1 - 2 287 93
GenBank mRNA AK013504.1 - 1 - 2 187 93
AK135036.1 - 1 - 2 179 93
AY566830.1 - 1 - 2 88 93
BC050763.1 - 1 - 2 171 93
BC080860.1 - 1 - 2 128 93
BY127313.1 - 1 - 2 288 93
U64572.1 - 1 - 2 147 93

View Full Product Details