The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010837.3 | NP_034967.2 | 3 - 4 | 2281 | 75 |
GenBank mRNA | AK018193.2 | - | 3 - 4 | 1761 | 75 |