The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166381.1 | NP_001159853.1 | 5 - 6 | 486 | 64 |
NM_001166382.1 | NP_001159854.1 | 5 - 6 | 486 | 64 | |
NM_001166383.1 | NP_001159855.1 | 5 - 6 | 486 | 64 | |
NM_011236.2 | NP_035366.2 | 5 - 6 | 486 | 64 | |
GenBank mRNA | AK080702.1 | - | 5 - 6 | 459 | 64 |
BC010673.1 | - | 4 - 5 | 358 | 64 | |
BC019123.1 | - | 5 - 6 | 466 | 64 | |
CJ057181.1 | - | 3 - 4 | 299 | 64 | |
U06837.1 | - | 5 - 6 | 408 | 64 | |
U12135.1 | - | 5 - 6 | 455 | 64 | |
Z32767.1 | - | 5 - 6 | 480 | 64 |