Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Thbs1
Entrez Gene ID
21825
Gene Name
thrombospondin 1
Gene Aliases
TSP-1, TSP1, Thbs-1, tbsp1
Chromosome Location
Chr.2: 118111890 - 118127133 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011580.3 NP_035710.2 10 - 11 1836 80
GenBank mRNA AK080686.1 - 10 - 11 1871 80
AK145202.1 - 10 - 11 1866 80
AK163092.1 - 10 - 11 1863 80
BC042422.1 - 10 - 11 1838 80
BC050917.1 - 10 - 11 1836 80
M87276.1 - 10 - 11 1862 80

View Full Product Details