The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166249.1 | NP_001159721.1 | 2 - 3 | 308 | 78 |
NM_001166250.1 | NP_001159722.1 | 2 - 3 | 623 | 78 | |
NM_001166251.1 | NP_001159723.1 | 2 - 3 | 623 | 78 | |
NM_011844.4 | NP_035974.1 | 2 - 3 | 308 | 78 | |
GenBank mRNA | AJ001118.2 | - | 2 - 3 | 236 | 78 |
AJ316580.1 | - | 2 - 3 | 499 | 78 | |
AK006949.1 | - | 2 - 3 | 619 | 78 | |
AK028008.1 | - | 2 - 3 | 251 | 78 | |
AK131645.1 | - | 2 - 3 | 288 | 78 | |
AK148242.1 | - | 2 - 3 | 251 | 78 | |
AK148395.1 | - | 2 - 3 | 251 | 78 | |
AK182184.1 | - | 1 - 2 | 151 | 78 | |
AK185666.1 | - | 2 - 3 | 173 | 78 | |
AK188276.1 | - | 1 - 2 | 151 | 78 | |
AK190239.1 | - | 1 - 2 | 151 | 78 | |
AK196912.1 | - | 1 - 2 | 151 | 78 | |
AK197275.1 | - | 1 - 2 | 151 | 78 | |
AK198283.1 | - | 1 - 2 | 151 | 78 | |
AK198733.1 | - | 1 - 2 | 151 | 78 | |
AK206822.1 | - | 2 - 3 | 173 | 78 | |
AK208240.1 | - | 2 - 3 | 173 | 78 | |
AK211363.1 | - | 2 - 3 | 173 | 78 | |
AK215869.1 | - | 2 - 3 | 218 | 78 | |
BC057965.1 | - | 2 - 3 | 233 | 78 | |
BY273849.1 | - | 2 - 3 | 81 | ||
BY706980.1 | - | 2 - 3 | 627 | 78 |