The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011960.2 | NP_036090.2 | 3 - 4 | 1523 | 148 |
GenBank mRNA | AF079557.1 | - | 3 - 4 | 1272 | 148 |
AK036656.1 | - | 3 - 4 | 1546 | 148 | |
AK160742.1 | - | 2 - 3 | 997 | 148 | |
AK164013.1 | - | 3 - 4 | 1510 | 148 | |
BC050892.1 | - | 3 - 4 | 1522 | 148 | |
BC053037.1 | - | 3 - 4 | 1506 | 148 | |
BC059827.1 | - | 3 - 4 | 1532 | 148 |