The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163588.1 | NP_001157060.1 | 2 - 3 | 650 | 69 |
NM_001163589.1 | NP_001157061.1 | 2 - 3 | 650 | 69 | |
NM_013639.2 | NP_038667.2 | 2 - 3 | 650 | 69 | |
GenBank mRNA | AK149021.1 | - | 2 - 3 | 652 | 69 |
BC046291.1 | - | 2 - 3 | 643 | 69 | |
X15475.1 | - | 2 - 3 | 412 | 69 |