Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prph
Entrez Gene ID
19132
Gene Name
peripherin
Gene Aliases
Prph1
Chromosome Location
Chr.15: 99053575 - 99058978 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163588.1 NP_001157060.1 5 - 6 1360 94
NM_001163589.1 NP_001157061.1 6 - 7 1264 94
NM_013639.2 NP_038667.2 5 - 6 1360 94
GenBank mRNA AK149021.1 - 5 - 6 1363 94
BC046291.1 - 6 - 7 1257 94
CB723542.1 - 1 - 2 108 94
X15475.1 - 5 - 6 1122 94

View Full Product Details