Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mkln1
Entrez Gene ID
27418
Gene Name
muskelin 1, intracellular mediator containing kelch motifs
Gene Aliases
A130067F06Rik, AU015903, BB154892
Chromosome Location
Chr.6: 31398828 - 31511795 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013791.2 NP_038819.1 12 - 13 1557 70
GenBank mRNA AK077541.1 - 12 - 13 1552 70
AK136123.1 - 9 - 10 1242 70
BC013703.1 - 12 - 13 1541 70
U72194.1 - 12 - 13 1540 70

View Full Product Details