The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016891.3 | NP_058587.1 | 2 - 3 | 220 | 77 |
GenBank mRNA | AB021743.1 | - | 2 - 3 | 168 | 77 |
AK054239.1 | - | 2 - 3 | 180 | 77 | |
AK078135.1 | - | 2 - 3 | 199 | 77 | |
AK088776.1 | - | 2 - 3 | 192 | 77 | |
BC006606.1 | - | 2 - 3 | 220 | 77 | |
BC052678.1 | - | 2 - 3 | 216 | 77 |