The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016910.3 | NP_058606.3 | 3 - 4 | 1023 | 78 |
GenBank mRNA | AF200464.1 | - | 3 - 4 | 804 | 78 |
AK157818.1 | - | 3 - 4 | 1011 | 78 | |
AK171714.1 | - | 3 - 4 | 1003 | 78 | |
BC023492.1 | - | 3 - 4 | 899 | 78 | |
BC051966.1 | - | 3 - 4 | 1023 | 78 |