The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016985.2 | NP_058681.1 | 14 - 15 | 2005 | 109 |
GenBank mRNA | AF073997.1 | - | 14 - 15 | 1978 | 109 |
AK048714.1 | - | 6 - 7 | 1258 | 109 | |
AK051390.1 | - | 7 - 8 | 1003 | 109 | |
AK082753.1 | - | 2 - 3 | 206 | 109 | |
BC056376.1 | - | 14 - 15 | 2005 | 109 | |
BC057337.1 | - | 14 - 15 | 1999 | 109 |