The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018822.3 | NP_061292.2 | 1 - 2 | 117 | 74 |
GenBank mRNA | AF153827.1 | - | 1 - 2 | 91 | 74 |
AF304053.1 | - | 1 - 2 | 91 | 74 | |
AK052266.1 | - | 1 - 2 | 110 | 74 | |
AK076267.1 | - | 1 - 2 | 146 | 74 | |
AK128926.1 | - | 1 - 2 | 117 | 74 | |
AK141266.1 | - | 1 - 2 | 65 | 74 | |
BC130268.1 | - | 1 - 2 | 114 | 74 |