Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdlim4
Entrez Gene ID
30794
Gene Name
PDZ and LIM domain 4
Gene Aliases
Ril
Chromosome Location
Chr.11: 54054928 - 54069032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019417.3 NP_062290.2 4 - 5 591 52
GenBank mRNA AB221672.1 - 4 - 5 506 52
BC030068.1 - 4 - 5 520 52
Y08361.1 - 4 - 5 542 52

View Full Product Details