Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pdlim4
Entrez Gene ID
30794
Gene Name
PDZ and LIM domain 4
Gene Aliases
Ril
Chromosome Location
Chr.11: 54054928 - 54069032 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019417.3 NP_062290.2 5 - 6 758 80
GenBank mRNA AB221672.1 - 5 - 6 673 80
AI385555.1 - 2 - 3 80
BC030068.1 - 5 - 6 687 80
Y08361.1 - 5 - 6 709 80

View Full Product Details