Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkp1
Entrez Gene ID
18772
Gene Name
plakophilin 1
Gene Aliases
AI528551
Chromosome Location
Chr.1: 135871395 - 135919361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019645.2 NP_062619.1 4 - 5 859 95
GenBank mRNA AK036856.1 - 4 - 5 1044 95
AK079429.1 - 4 - 5 1044 95
BC139041.1 - 4 - 5 954 95
BC139042.1 - 4 - 5 955 95
Y07941.1 - 4 - 5 859 95

View Full Product Details