The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170907.1 | NP_001164378.1 | 9 - 10 | 1501 | 67 |
NM_019797.4 | NP_062771.2 | 12 - 13 | 2317 | 67 | |
GenBank mRNA | AF197574.1 | - | 9 - 10 | 1376 | 67 |
AF539614.1 | - | 12 - 13 | 2200 | 67 | |
AK039024.1 | - | 9 - 10 | 1386 | 67 | |
AK078393.1 | - | 9 - 10 | 1378 | 67 |