Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rps6ka4
Entrez Gene ID
56613
Gene Name
ribosomal protein S6 kinase, polypeptide 4
Gene Aliases
1110069D02Rik, 90kDa, AI848992, Msk2, mMSK2
Chromosome Location
Chr.19: 6829080 - 6841045 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019924.2 NP_064308.2 1 - 2 135 58
GenBank mRNA AF074714.1 - 1 - 2 107 58
AK089467.1 - 1 - 2 127 58
AK154674.1 - 1 - 2 134 58
BC012964.1 - 1 - 2 145 58
CJ065157.1 - 1 - 2 130 58

View Full Product Details